Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "longsleeve-shirts"
SearchPresentationType-VERTICAL
SearchResultCount:"11"
  List View Grid View Easy View (new)
Sort by:
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   These soft shirts are made of Ultra Line double knitted fabric: The exterior is made of 100% polyester, the interior is made of 50% Viloft® + 50% poly...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Soft shirt made of 60% Protex® and 40% Viloft® (single jersey). The inherently flame-retardant fabric has good moisture regulating properties and feat...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Soft shirt made of 60% Protex® and 40% Viloft® (single jersey). The inherently flame-retardant fabric has good moisture regulating properties and feat...
Supplier:  Portwest Clothing
Description:   Shirts with long sleeves made of 50% polyester and 50% cotton. The fabric offers warmth at all times.
Supplier:  Portwest Clothing
Description:   Baker shirt made of fully pre-shrunk, heavyweight poly cotton fabric (TradeGuard), hot head pressed.
Supplier:  FRISTADS KANSAS GROUP
Description:   Shirt made of 100% spunbounded cotton (single jersey), with anti-pilling treatment. The grey melange version is manufactured from 85% cotton and 15% v...
Supplier:  FRISTADS KANSAS GROUP
Description:   ESD shirt with long sleeves, manufactured from a knitted blend (83% cotton, 14% polyester and 3% conductive fibre), which makes it comfortable to wear....
Supplier:  FRISTADS KANSAS GROUP
Description:   Long sleeved shirts made of 44,5% polyester (Cocona®) and 55,5% standard polyester. Cocona® fabric transports moisture, absorbs odour, dries very quic...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 11
  1  2  Next