Separating funnels, cylindrical
Supplier: Hecht
HECH42956030EA 43.3 EUR
HECH42956030
Separating funnels, cylindrical
Separating funnel, cylindrical, made of borosilicate glass 3.3, with solid glass plug with glass key and plastic stopper.
  • With inscription label
  • Excellent chemical
  • Excellent temperature resistance