Test tubes with standard ground stoppers
Supplier: Hecht
HECH42767030EA 32 EUR
HECH42767030 HECH42766010 HECH42768002
Test tubes with standard ground stoppers
Tubes Test Tubes
AR-Glas® (soda glass).
  • Ungraduated or graduated or with ringmarks and numbers
  • Good chemical and physical properties
Round bottom, with standard ground and stopper of polyethylene (HDPE).