You Searched For: Specialty Cuvettes
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (634-0944)
Supplier: HELLMA
Description: Optical glass cuvette designed for measuring amplitudes of turbidity.
UOM: 1 * 1 items
Catalog Number: (KART2618)
Supplier: KARTELL
Description: These cylindrical cells are specifically used in conjunction with Olli-C-Analyzer.
UOM: 1 * 2.688 items
Catalog Number: (TINT605534)
Supplier: Lovibond Tintometer
Description: Absorptiometer cell made of optical glass with path length of 10 mm.
UOM: 1 * 10 items
Catalog Number: (KART2630)
Supplier: KARTELL
Description: Polystyrene cuvettes for Fibrintimer used to determine the coagulation-time of blood.
UOM: 1 * 500 items
Supplier: KARTELL
Description: Polystyrene cuvettes designed for the determination of coagulation time.
Supplier: KARTELL
Description: These twelve position cuvettes segments have been specially designed for use with the Cobas Mira® analyser.
Catalog Number: (PERCB0017054)
Supplier: PerkinElmer
Description: Glass cells designed for easy filling and emptying with no loss of sample.
UOM: 1 * 1 items
Catalog Number: (HACHHBK059)
Supplier: Hach
Description: UV flow through cuvette for Ganimede N nitrogen analyser.
UOM: 1 * 1 items
Catalog Number: (LPIT110511)
Supplier: LP ITALIANA SPA
Description: Cuvettes for Sherwood coagulometer.
UOM: 1 * 1.000 items
Catalog Number: (SARS68.752)
Supplier: SARSTEDT
Description: Polypropylene round opaque cuvette for sample preparation with LKB analyser.
UOM: 1 * 5.000 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom