Certificates
Chemical Structure
Manufacturer Data
Login
/
Register
Slovenia
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
Proteins and Peptides
E-64C
E-64C
E-64C
Supplier:
Cayman Chemical
Certificates
Expand image
E-64C
Supplier:
Cayman Chemical
About this item
Specifications
Source:
Species:
Environmentally Preferable:
Specifications
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
CAYM10007964-10
10007964-10
Glass bottle for solids
10 mg
Log in
for availability
Log in
for availability
1 * 10 mg
1 * 10 mg - 633,00 €
633,00 €
New Customer Promotional Price
-
+
Add to cart
1 * 10 mg
633,00 €
-
+
Add to cart
CAYM10007964-25
10007964-25
Glass bottle for solids
25 mg
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 1 380,00 €
1 380,00 €
New Customer Promotional Price
-
+
Add to cart
1 * 25 mg
1 380,00 €
-
+
Add to cart