Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Pullovers"
https://si-bot.vwr.com/stibo/smallweb/std.lang.all/96/92/18989692.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"12"
  List View Grid View Easy View (new)
Sort by:
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Soft pullover made of 71% molleton polyester, 25% viscose and 4% elasthane. With decorative seams and a grey finish at cuffs and collar.
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Pullover made of invisible rugged molton with w-binding. The soft fabric consists of 48% Polyester, 48% Viloft and 4% elasthane. It provides good ther...
Supplier:  Portwest Clothing
Description:   This classic style sweatshirt is made of 50% polyester and 50% cotton.
Supplier:  Portwest Clothing
Description:   The comfortable B300 Roma sweatshirt is ideal for work wear or casual weekend wear. Made of 65% polyester and 35% cotton.
Supplier:  Portwest Clothing
Description:   These sweathshirts are made from a special blend of 65% polyester and 35% cotton with a brushed back for extra warmth and comfort. The fabric provides...
Supplier:  FRISTADS KANSAS GROUP
Description:   Sweatshirt with crew neck, made from 80% cotton and 20% polyester.
Supplier:  FRISTADS KANSAS GROUP
Description:   Army-style sweatshirt made of 80% wool and 20% polyamide. Ideal for service industries.
Supplier:  FRISTADS KANSAS GROUP
Description:   Sweatshirt made of 50% polyester and 50% cotton. With reflective tape for improved visibility.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 12
  1  2  Next