Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "outdoor-caps-and-headbands"
SearchPresentationType-VERTICAL
SearchResultCount:"15"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Portwest Clothing
Description:   This classic fleece hat made of 100% polyester fleece is ideal for outdoor use.
Supplier:  Portwest Clothing
Description:   White unisex nylon mesh hat. Ideal for use in the food industry.
Catalog Number: (113-8804)
Supplier:  Portwest Clothing
Description:   This reversible beanie is made from 100% polyester fleece. It is ideal for use in areas where high visibility is required.
UOM:  1 * 1 items
Supplier:  COTTON CLASSICS HANDELS GMBH
Description:   6 panel cap with rip-tape fastening, made from 100% heavy non brushed cotton.
Supplier:  FELDTMANN HELMUT KG
Description:   Polyester/cotton, Hi-vis orange/grey
Supplier:  COTTON CLASSICS HANDELS GMBH
Description:   Myrtle Beach®, MB 7551, Acrylic, Dark green, 70 g
Supplier:  FRISTADS KANSAS GROUP
Description:   Knitted cap made of 100% acrylic.
Catalog Number: (113-6407)
Supplier:  FRISTADS KANSAS GROUP
Description:   Double knitted beanie made of 100% acrylic.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 15
  1  2  Next