Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (3)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "industrial-overalls"
SearchPresentationType-VERTICAL
SearchResultCount:"55"
  List View Grid View Easy View (new)
Sort by:
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Water- and windproof overalls made of Flexothane® Classic (100% polyamide knitting with PU coating). With fixed quilted lining for improved cold prote...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Lightweight, durable overall made of Flexothane® Classic fabric consisting of 100% polyamide knitting with PU coating.
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Lightweight, durable overall made of Flexothane® Classic fabric consisting of 100% polyamide knitting with PU coating. The fabricl is water- and windp...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Water- and windproof overall made of Flexothane® Classic: 100% polyamide knitting with PU coating. With fixed quilted lining, knitted wind cuffs in sl...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Water- and windproof overalls made of Flexothane Classic, 100% polyamide knitting with PU coating.
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   These unlined overalls are made of Flexothane® Flame fabric consisting of 100% FR polyester knitting with FR PU coating. With high frequency welded se...
Supplier:  DuPont
Description:   The Tyvek® Dual garment combines Tyvek® on the front with a non woven SMS panel on the back. The Tyvek® front is permeable to both air and water vapou...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   These water- and windproof overalls are made of Siopor® FR AST, a polyester fabric with 100% FR PU coating + AST. The fabric is highly breathable, wat...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 55
  1  2  3  4  5  6  7  Next