Magnetic stirring bars, Octoval
COWI001.3376EA 62.4 EUR
COWI001.3376 COWI001.3364 COWI001.3341 COWI001.3325 COWI001.3319 COWI001.3338 COWI001.3351
Magnetic stirring bars, Octoval
PTFE covered. Effective magnetic stirrers with similar action to oval/privot ring types.
  • Very good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)