NIR polarimetric saccharimeters, ADS480
Supplier: Bellingham + Stanley
635-0801EA 0 EUR
635-0801 635-0802 635-0799 635-0800 635-0797 635-0798 635-0795 635-0796 635-0793 635-0794 635-0803 635-0804
NIR polarimetric saccharimeters, ADS480
Polarimeters
ASDS480 is a Near Infra Red (NIR) automatic polarimeter designed for laboratories and tare houses, especially for the cane, beet and ethanol industries.
  • High quality optics for transmissions in the infrared spectrum
  • Sensor providing the measurement for sugar or quartz temperature compensation
  • Calibration using a quartz control plate by accessing the span facility in the set-up menu
PC software for 'purity' calculation with a refractometer and touch sensitive LCD screen for excellent robustness in tares houses.
Certifikace: ICUMSA
Upozornení: For the sugar and associated industries, where lead acetate is not allowed, therefore dictating the need for an infrared measurement.