You Searched For: Enzymatic Tests
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (941970ZA)
Supplier: VWR Chemicals
Description: The catalase test can differentiate or identify microorganisms by checking their ability to produce catalase, an enzyme that protects microorganisms f...
UOM: 1 * 30 mL
Catalog Number: (942152ZF)
Supplier: VWR Chemicals
Description: Swabs imprenated with reagent for the detection and verification of bacterial cytochrome oxidase.
UOM: 1 * 50 Tests
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: RapIDâ„¢ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapIDâ„¢ panels offer easy, on...
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Enzymatic Tests, Microbactâ„¢ Reagent Indole-Kovacs
Supplier: LIOFILCHEM S.R.L.
Description: EnteroPluri Test is a 12-sector system containing special culture media that permits identification of the Enterobacteriaceae and other gram ne...
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: The Oxoid Biochemical Identification System (O.B.I.S.) is a rapid colorimetric test for the determination of biochemical activity in microorganisms. I...
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Enzymatic Tests, Pathotec CO strips
Catalog Number: (OXOIMB1131A)
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: The Microbactâ„¢ Gram-negative system is to be used for the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteria...
UOM: 1 * 40 Tests
Supplier: Hach
Description: Biological Activity Reaction Test (BART) biodetectors are microbiological diagnostic tools used to identify the presence of process disruptors. Tests ...
Catalog Number: (APPC416444.2326)
Supplier: PanReac AppliChem
Description: Plastic sticks used for the easy and rapid detection of the cytochrome oxidase enzyme in microbiological diagnosis. Oxidase sticks have a much higher ...
UOM: 1 * 50 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom