Proteinase K
E195-5MLEA 124 EUR
VWRCE195-5ML
Proteinase K
A non specific serine protease with a molecular weight of approximately 18 kDa. This enzyme exhibits high stability and activity in the presence of SDS, EDTA and urea, as well as over a wide pH range. Activity >30 units/mg.
Proteinase K is useful for the inactivation of nucleases during the isolation of DNA and RNA. Active over a broad pH range.