You Searched For: Food Standards
61  results were found
SearchResultCount:"61"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (1087494.)
Supplier: USP
Description: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
UOM: 1 * 500 mg
Catalog Number: (1269119.)
Supplier: USP
Description: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
UOM: 1 * 100 mg
Catalog Number: (1268919.)
Supplier: USP
Description: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
UOM: 1 * 1 g
Catalog Number: (1036959.)
Supplier: USP
Description: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
UOM: 1 * 1 mL
Catalog Number: (1294839.)
Supplier: USP
Description: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
UOM: 1 * 350 g
Catalog Number: (1548098.)
Supplier: USP
Description: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
UOM: 1 * 5 mL
Catalog Number: (1036960.)
Supplier: USP
Description: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
UOM: 1 * 500 mg
Catalog Number: (CAYM29557-5)
Supplier: Cayman Chemical
Description: BSA-Oleate monounsaturated fatty acid complex is composed of oleic acid and bovine serum albumin (BSA) at an approximately 6:1 molar ratio of oleate:B...
UOM: 1 * 5 mL
Catalog Number: (CAYM29558-5)
Supplier: Cayman Chemical
Description: BSA-Palmitate saturated fatty acid complex is composed of palmitic acid and bovine serum albumin (BSA) at an approximately 6:1 molar ratio of palmita...
UOM: 1 * 5 mL
Catalog Number: (REST35025)
Supplier: Restek
Description: This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like olive, teaseed, and neat...
UOM: 1 * 1 mL
Catalog Number: (REST32562)
Supplier: Restek
Description: Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea...
UOM: 1 * 1 Each
Supplier: Restek
Description: Methyl caprate
Catalog Number: (EHERC12972364)
Supplier: EHRENSTORFER GMBH
Description: Disperse yellow 54, Dr Ehrenstorfer
UOM: 1 * 25 mg
Catalog Number: (REST35023)
Supplier: Restek
Description: This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like linseed, perilla, hempse...
UOM: 1 * 1 items
Supplier: Restek
Description: These standards are used for pesticide residue quantification.
Supplier: Agilent
Description: The Organic Acids Solutions Kit contains capillaries, buffers, an organic acids standard and instructions complete with a sample electropherogram to h...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 61
no targeter for Bottom