You Searched For: Precast Gels
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: EDVOTEK EUROPE LTD
Description: Precast gel, 6 gels (12%)
Supplier: LONZA
Description: Latitudeâ„¢ DNA gels are designed for accurate DNA fragment separation for almost any gel electrophoresis application. These ready-to-run gels are avail...
Supplier: LONZA
Description: Reliantâ„¢ precast agarose minigels are designed for rapid and reproducible resolution of DNA fragments from 8 bp to 10 kb. Each precast agarose gel is ...
Supplier: LONZA
Description: Precast gels for the analysis of antibodies and proteins up to 2000 kDa.
Supplier: G-Biosciences
Description: This gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permane...
Catalog Number: (28-9374-51)
Supplier: Cytiva
Description: DIGE gel is a 12,5% precast polyacrylamide gel in a low fluorescent glass cassette for second dimension 2-D electrophoresis and specially developed fo...
UOM: 1 * 3 items
Supplier: Cytiva
Description: Immobilineâ„¢ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate app...
Supplier: Cytiva
Description: ExcelGel precast gels for SDS-PAGE are available as homogeneous or gradient gels. The homogeneous gels have 25 preformed sample wells for sample volum...
Supplier: Genscript
Description: GenScript's Bis-Tris gels are high-performing polyacrylamide gels designed to separate a wide range of protein sizes by electrophoresis. The gels are ...
Supplier: EMBITEC
Description: RunOneâ„¢ precast gels do not require a specific gel apparatus but are optimised for use with the RunOneâ„¢ Electrophoresis System. A choice of 100, 104 o...
Supplier: Cytiva
Description: Precast gel, PhastGel™, PhastGel™ sample applicator, 8×0,5 μl
Sale
Catalog Number: (18-1035-32)
Supplier: Cytiva
Description: CleanGel IEF is a dried polyacrylamide gel optimised for analytical isoelectric focusing (IEF). The gel is excellent for separating enzymes with prese...
UOM: 1 * 5 items
MSDS
Catalog Number: (80-1128-28)
Supplier: Cytiva
Description: Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients.
UOM: 1 * 3 items
Supplier: Serva
Description: For cerebrospinal fluid (CSF) analysis by isoelectric focusing.
Catalog Number: (SERA42803.01)
Supplier: Serva
Description: For transferrin (Tf) analysis by isoelectric focusing.
UOM: 1 * 1 items
Catalog Number: (SERA42805.01)
Supplier: Serva
Description: For haptoglobin (Hp) analysis by isoelectric focusing.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 17
no targeter for Bottom