You Searched For: Electrode Filling Solutions
29  results were found
SearchResultCount:"29"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Potassium chloride 3 Molar solution may also be used as an electrode storage solution.
Catalog Number: (662-1116)
Supplier: VWR Collection
Description: Calomel-free pH electrode refill solution containing potassium nitrate , VWR®
UOM: 1 * 114 mL
MSDS
Catalog Number: (662-1784)
Supplier: VWR Collection
Description: Refill solution 3,5 mol/l KCl, VWR®
UOM: 1 * 100 mL
MSDS
Supplier: Hach
Description: Filling and storage solutions for combined and reference electrodes.
Supplier: Hach
Description: Electrolyte solution 0,1 M, 125 ml, for sensION+ ISE for sodium ion in range 0,023 - 23000 mg/lL
Supplier: HAMILTON BONADUZ
Description: A variety of electrolyte solutions for pH and ORP electrodes.
Supplier: Hach
Description: Electrolyte solution (KCl 3M), 125 ml
Catalog Number: (HACH28564026)
Supplier: Hach
Description: Filling solution for electrode reference element, potassium nitrate (KNO₃), 1 M solution.
UOM: 1 * 59 mL
Supplier: Hach
Description: Reagents in Powder Pillow form - typically as powder which does not appreciably affect the dilution of the sample.
Supplier: Hach
Description: Shelf life: 48 months from production date.
Catalog Number: (HACH2841700)
Supplier: Hach
Description: Internal filling solution for IntelliCALâ„¢ PHC301 pH probes and MTC301 ORP probes, 3,0M KCl, saturated with silver chloride (AgCl), 28 ml bottle
UOM: 1 * 28 mL
Catalog Number: (HACH2824349)
Supplier: Hach
Description: Ammonia Ionic Strength Adjustor (ISA) buffer solution for ammonia determinations by electrode methods.
UOM: 1 * 500 mL
Supplier: Hach
Description: Electrolyte filling solution (25 ml) for Sension+ 5120 dissolved oxygen probe
Catalog Number: (HACH2965026)
Supplier: Hach
Description: Internal filling solution for use with PHC281 IntelliCALâ„¢ Ultra pH probes, 2,44M KCl, 59 ml bottle with dispenser cap, IntelliCALâ„¢
UOM: 1 * 59 mL
Catalog Number: (HACH5194100)
Supplier: Hach
Description: pH electrode filling solution, 4M KCl saturated with silver chloride (AgCl ), 65 ml
UOM: 1 * 65 mL
Supplier: Mettler - Toledo
Description: FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 29
no targeter for Bottom