Order Entry
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for AAS
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for AAS
Supplier:  AccuStandard
Certificates
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid standard for AAS
Supplier:  AccuStandard
Specifications
  • Environmentally Preferable:
Specifications
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • ACCUAA13N5
    AA13N5
    Chromium
    Cr
    1000 mg/L
    Nitric acid dil.
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL
  • ACCUAA13N1
    AA13N1
    Chromium
    Cr
    1000 mg/L
    Nitric acid dil.
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL