You Searched For: Slide Holders
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Slide holders designed for applications in clinical cytology.
Supplier: VWR Collection
Description: Slide trays hold 20 or 30 standard microscopy slides (76×26 mm).
Supplier: VWR Collection
Description: Q Path® Quickspot and slide holder for Turbitec are used to carry out cytocentrifugation. They allow a cell suspension to be deposited on a slide (gyn...
Supplier: VWR Collection
Description: Flip-top trays designed for the safe storage of up to 20 microscope slides. With or without lid.
Sale
Catalog Number: (631-1049)
Supplier: VWR Collection
Description: This metallic cabinet is suitable for the storage of 50×50 mm or 80×57 mm microscopy slides.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Sturdy microscope slide holder designed for drying and cleaning.
Supplier: VWR Collection
Description: These organisers enable the archiving of a large number of microscope slides.
Catalog Number: (631-0823)
Supplier: VWR Collection
Description: Cardboard microscope slide holders offer instant reference and fast, fingertip retrieval.
UOM: 1 * 12 items
Catalog Number: (631-1064)
Supplier: VWR Collection
Description: Slide cabinet for long-term storage.
UOM: 1 * 10 items
Catalog Number: (631-0835)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Slide dispenser, Grey, No. of slides: 72
UOM: 1 * 1 items
Supplier: Simport Scientific
Description: The SlideFolder is manufactured from heat-resistant, injection moulded plastic and can hold up to 20 standard microscope slides.
Supplier: Simport Scientific
Description: This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individually numbered slots. All slides are stored upright for easie...
Catalog Number: (720-0487)
Supplier: Simport Scientific
Description: This drawer is made from heat resistant injection moulded plastic.
UOM: 1 * 10 items
Catalog Number: (LEIH3808500E)
Supplier: LEICA HISTOLOGY
Description: The slide cabinet is a storage system that can hold up to 6430 standard microscope slides.
UOM: 1 * 1 items
Catalog Number: (630-2771)
Supplier: MARIENFELD
Description: Preparation table for 20 microscopy slides. Thanks to its recessed grips the slides can be easily taken out.
UOM: 1 * 20 items
Catalog Number: (LEIH3808587E)
Supplier: LEICA HISTOLOGY
Description: This drawer cabinet includes eight drawers and can be stacked for comfortable use.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom