Accessories for homogeniser, HO 4/A
- Pk
- Knife, stainless steel, for 150 ml vessels
Specifications
Environmentally Preferable - Sample vessel, 25 ml, DURAN® borosilicate glass
Specifications
Environmentally Preferable - Sample vessel, 10 ml, DURAN® borosilicate glass
Specifications
Environmentally Preferable - Knife, stainless steel, for 25 ml vessels
Specifications
Environmentally Preferable - Knife, stainless steel, for 10 ml vessels
Specifications
Environmentally Preferable - Knife, stainless steel, for 100 ml vessels
Specifications
Environmentally Preferable - Knife, stainless steel, for 50 ml vessels
Specifications
Environmentally Preferable - Sample vessel, 100 ml, DURAN® borosilicate glass
Specifications
Environmentally Preferable - Sample vessel, 50 ml, DURAN® borosilicate glass
Specifications
Environmentally Preferable - Spindle, stainless steel, for 25 - 150 ml vessels
Specifications
Environmentally Preferable - Spindle, stainless steel, for 10 ml vessels
Specifications
Environmentally Preferable - Sample vessel, 150 ml, DURAN® borosilicate glass