You Searched For: Microscope Slides
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Microscopy slides are designed with colour-frosted ends for H&E stain on frozen sections in histopathology examination.
Supplier: VWR Collection
Description: Colour frosted microscopy slide, Bevelled edges, 45°, VWR®, White
Sale
Supplier: KLINIPATH
Description: Microscope slides for slide printers allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
Sale
Supplier: VWR Collection
Description: Microscopy slides are designed with colour-frosted ends for H&E stain on frozen sections in histopathology examination.
Supplier: VWR Collection
Description: These microscope slides come with a light opaque coating in different colours.
Supplier: VWR Collection
Description: Microscope slides, VWR®, Edges: Cut, L×W: 76×26 mm, Thickness: 1,0 mm
Catalog Number: (631-1161)
Supplier: VWR Collection
Description: Microscope slides with double-frosted ends designed for human blood, body fluids tests, as well as paraffin sections.
UOM: 1 * 2.500 items
Catalog Number: (631-1160)
Supplier: VWR Collection
Description: Microscope slides with ground edges designed for human blood or fluid tests.
UOM: 1 * 2.500 items
Catalog Number: (KLINPR-001)
Supplier: KLINIPATH
Description: Microscope slides specially designed for use with printers.
UOM: 1 * 2.500 items
Supplier: Epredia
Description: These slides are made of extra-white glass, washed clean and polished.
Supplier: MARIENFELD
Description: Slides designed for in vitro diagnostic (IVD) applications.
Sale
Supplier: Corning
Description: These slides are either frosted on one side, on both sides, or plain.
Supplier: Epredia
Description: These microscope slides are available plain or with one frosted end.
Catalog Number: (MARI0704022)
Supplier: MARIENFELD
Description: UniMark slide is designed with a white circle of Ø 12 mm and a white marking area of 20 mm for use in cytocentrifuges.
UOM: 1 * 2.500 items
Catalog Number: (MARI1527120091)
Supplier: MARIENFELD
Description: Microscopy slides with chamfered corners to reduce the risk of injuries.
UOM: 1 * 25 items
Supplier: Epredia
Description: These slides come with a light opaque coating in standard colours.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom