Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "titrants"
https://si.vwr.com/stibo/smallweb/std.lang.all/99/22/18989922.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"20"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Chemicals
Description:   These reagents are especially suitable for water determination in aldehydes and ketones.
Supplier:  VWR Chemicals
Description:   Both anolyte and catholyte solutions are with improved formulation to increase the speed and accuracy of titration when determining water content at t...
Supplier:  VWR Chemicals
Description:   Universal indicator ethanol solution TECHNICAL®
UOM:  1 * 1 l
Supplier:  Alfa Aesar
Description:   Karl Fischer Coulometric CG, catholyte solution 1 * 6 items
UOM:  1 * 6 items
Supplier:  Hach
Description:   For determination of low level manganese by the PAN method.
Supplier:  Hach
Description:   Wide range of pH indicator solutions used for visual determination of pH, using test kits.
Catalog Number: (HACH2308232)
Supplier:  Hach
Description:   This titrant is used for the determination of ascorbic acid in juice based beverages, by drop-count titration.
UOM:  1 * 100 ml
Catalog Number: (HACH2589636)
Supplier:  Hach
Description:   These reagents are used for determining ultra-low hardness by the chlorophosphonazo method.
UOM:  1 * 10 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 20
  1  2  3  Next