You Searched For: Colour Reference and Spectroscopy Standards
37  results were found
SearchResultCount:"37"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: These solutions are developed and produced according to chapter 2.2.2. 'Degree of colouration of liquids' of the European Pharmacopoeia. (brown, brown...
Supplier: VWR Chemicals
Description: For spectrophotometer calibration according to European Pharmacopoeia (Ph. Eur.)
Supplier: VWR Chemicals
Description: Complete range of reagents and standard solutions in Chapters 2 and 4 of the current European Pharmacopoeia.
All are manufactured and tested in c...
Catalog Number: (H296-03)
Supplier: MACRON (A AVANTOR BRAND)
Description: Corrosive.
UOM: 1 * 500 mL
Supplier: VWR Chemicals
Description: Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis.
Supplier: VWR Chemicals
Description: Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis that are designed for grading the colour of similarly coloure...
Supplier: VWR Chemicals
Description: Ready to use colour solutions according to Reag. Ph. Eur.
Supplier: VWR Chemicals
Description: Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis.
Supplier: VWR Chemicals
Description: Standard for spectrophotometer calibration, Holmium perchlorate standard solution, Reag. Ph. Eur, VWR®
Supplier: VWR Chemicals
Description: Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis and are designed to measure the visual colour of water suppli...
Catalog Number: (84833.180)
Supplier: VWR Chemicals
Description: Bandwidth standards.
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Wavelength standards (certified at 0,1; 0,2; 0,5; 1 and 2 nm slit widths) packed in six permanently sealed UV cuvettes or 100 ml amber bottles.
Catalog Number: (84756.180)
Supplier: VWR Chemicals
Description: Linearity standards packed in permanently sealed UV cuvettes or 100 ml amber bottles.
UOM: 1 * 100 mL
Catalog Number: (85969.001)
Supplier: VWR Chemicals
Description: A set for linearity check of the absorbance of spectrophotometers.
UOM: 1 * 4 parts
Supplier: VWR Chemicals
Description: Stray light standards packed in permanently sealed UV cuvettes or 100 ml amber bottle.
Catalog Number: (705-1122)
Supplier: Hach
Description: Calibration solutions for Lico spectral colorimeter.
UOM: 1 * 1 KIT
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 37
no targeter for Bottom