You Searched For: Cleanroom Wipes
173  results were found
SearchResultCount:"173"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Polyester wipe for use in ISO 4 cleanrooms.
Supplier: VWR Collection
Description: These wipes are made from 100% continuous filament, double-ply, ultrasonically-sealed polyester to provide better absorbency.
Supplier: VWR Collection
Description: Designed for cleaning laminar flow benches, cleanroom surfaces, quartzware and general process equipment. Appropriate for electronics, pharmaceutical,...
Catalog Number: (115-0033)
Supplier: VWR Collection
Description: These cleanroom wipes are made of polyester and cellulose.
UOM: 1 * 100 items
Supplier: VWR Collection
Description: Absorbent, non woven wipe for use in ISO 5 and 6 cleanrooms.
Supplier: VWR Collection
Description: Ideal for cleaning rough or uneven surfaces, yet still soft enough for use on sensitive surfaces. Use these 100% double-knit, polyester wipes where mi...
Supplier: VWR Collection
Description: These are 100% continuous-filament, double-knit polyester fabric wipes.
Catalog Number: (115-1830)
Supplier: VWR Collection
Description: Double ply polyester sealed-edge wipe for use in ISO 5 cleanrooms.
UOM: 1 * 50 items
Supplier: VWR Collection
Description: Microfibre, sealed-edge wipe for use in ISO 3 cleanrooms.
Catalog Number: (115-0034)
Supplier: VWR Collection
Description: These non woven wipes, made from 45% polyester and 55% cellulose, are designed for cleaning laminar flow benches, cleanroom surfaces and general proce...
UOM: 1 * 75 items
MSDS
Supplier: VWR Collection
Description: 2 ply polyester sealed-border wipe for use in ISO 3 cleanrooms.
Catalog Number: (115-1724)
Supplier: VWR Collection
Description: These wipes are made from double-woven polyester fabric, with close-cut edges, and are washed in an ISO Class 4 cleanroom to minimise contamination by...
UOM: 1 * 300 items
Supplier: VWR Collection
Description: Polyester, sealed-edge wipe for use in ISO 3 cleanrooms.
Supplier: VWR Collection
Description: Excellent for cleaning rough, abrasive, or irregular surfaces, yet soft enough for delicate surfaces. Appropriate for pharmaceutical and biotechnology...
MSDS Sale
Supplier: VWR Collection
Description: These wipes are made from 100% continuous filament, double-ply, ultrasonically-sealed polyester to provide better absorbency. The wipe's size is perfe...
Catalog Number: (115-1902)
Supplier: VWR Collection
Description: Polyester, sealed-border wipes for use in ISO 3 cleanrooms.
UOM: 1 * 150 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 173
no targeter for Bottom