You Searched For: Test/Sample Tube Closures
87  results were found
SearchResultCount:"87"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Safe-T-Flex caps feature a unique, flexible, over-locking design, locking securely on the outside of the tube lip and can be removed with one hand. A ...
Catalog Number: (525-0923)
Supplier: VWR Collection
Description: Purple PP. Sustainable design based on virgin resins.
UOM: 1 * 500 items
Low Manufacturing Impact
Sustainable Packaging
Supplier: VWR Collection
Description: Flat screw cap with O-ring for microcentrifuge tubes, VWR®, Colour: Red
Sale
Supplier: VWR Collection
Description: Cap for culture and test tube, PE, VWR®, Colour: Natural, Closure type: Dual position type, For tubes: 17 mm
Low Manufacturing Impact
Sustainable Packaging Sale
Supplier: VWR Collection
Description: These closures are suitable for test and culture tubes.
Catalog Number: (45000-0006)
Supplier: Avantor
Description: Strip of 8 plug caps, J.T.Baker®, Colour: Clear
UOM: 1 * 120 items
Supplier: VWR Collection
Description: Cap for culture and test tube, VWR®, Colour: Natural, Closure type: Dual position type, For tubes: 12 mm
Low Manufacturing Impact
Sustainable Packaging
Catalog Number: (634-0566)
Supplier: VWR Collection
Description: PTFE, white. Caps are machined to exact specifications with smaller gyroradius and more homogeneous mass distribution than disposable caps providing b...
UOM: 1 * 25 items
Supplier: Saint-Gobain
Description: Made of translucent silicone. Ideal for protecting piping/tubing or protruding cylindrical components during transport and all operations that require...
Sale
Supplier: SP Industries
Description: Manufactured in ethylene vinyl acetate (EVA).
Supplier: LUDI A.G.
Description: Aluminium.
Supplier: MATRIX TECHNOLOGIES CORP. LTD
Description: Caps for 0,5 ml and 1,0 ml screw top tubes, Matrix®, Colour: Purple
Supplier: Bohlender
Description: Red screw cap made of glass-fibre reinforced ETFE, inner parts consisting of a v-ring (ETFE), a tapered ring and a sealing ring (both PTFE) as well as...
Supplier: Simport Scientific
Description: Caps are available either with or without a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures.
Supplier: LUDI A.G.
Description: Made of anodised aluminum with stainless steel spring clip.
Supplier: SP Industries
Description: Made of PTFE, white. NMR tube caps are machined to exact specifications with smaller gyroradius and more homogeneous mass distribution than disposable...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 87
no targeter for Bottom