Ultrasonic baths, PC-620
Supplier: Branson
BRAN000-951-130EA 1470.88 EUR
BRAN000-951-130 BRAN000-951-430
Ultrasonic baths, PC-620
A bench-top cleaner specifically suited to cleaning long, narrow components such as pipettes, lab glassware and other items requiring general cleaning.