You Searched For: Swabs for Clinical Use
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: DELTALAB
Description: Swabs designed for cleaning and disinfection when tip is soaked in a disinfectant, thus reducing the risk of a direct contact.
Supplier: NISSUI PHARMA SOLUTIONS
Description: CompactDry Swabs for surfaces, meat, fish and poultry. New design with swabs pre immersed in 1 ml buffer, ready for use with a self-standing tube and ...
New Product
Catalog Number: (SCIL2200001)
Supplier: HYSERVE GMBH & CO. KG
Description: This Swab System is designed for sampling on surfaces in the production environment. It consists of a sterile swab in a tube with 10 ml sterile PBS bu...
UOM: 1 * 40 Tests
Catalog Number: (HERE1032018)
Supplier: HERENZ, HEINZ GMBH
Description: Wooden applicators with a double cotton bud.
UOM: 1 * 2.000 items
Catalog Number: (PMPC826-WC)
Supplier: PURITAN MEDICAL
Description: Tapered swabs designed for precision application and hard-to-reach areas.
UOM: 1 * 10.000 items
Catalog Number: (PMPC25-2199)
Supplier: PURITAN MEDICAL
Description: These cytology brushes feature tapered tips made out of soft nylon bristles and a long PS handle.
UOM: 1 * 1.000 items
Catalog Number: (SAFID3794)
Supplier: SAFETY FIRST AID GROUP LTD
Description: A first aid staple with numerous uses designed for cleaning and dressing wounds.
UOM: 1 * 100 items
Catalog Number: (HARV59-8422)
Supplier: HARVARD APPARATUS LTD
Description: These sterile cellulose sponges are designed for the removal of fluids during microsurgery.
UOM: 1 * 200 items
Supplier: HERENZ, HEINZ GMBH
Description: Aluminium applicators with a single cotton bud.
Catalog Number: (MURRL602/01)
Supplier: MURRAY, S & CO LTD
Description: Wooden applicator stick.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom