VWR®, Aprons, LDPE/LLDPE
Supplier: VWR
VWR®
113-8279EA 21.4 EUR
113-8279 113-8278
VWR®, Aprons, LDPE/LLDPE
Disposable Garments
This disposable apron is made from a mixture of 65% LDPE and 35% LLDPE.
  • Effective against dirt, moisture and oil
Length: 1200 or 1400 mm
Colour: White