Allyl chloroformate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2937-50-0
:
Formula:C₄H₅ClO₂
MDL Number:MFCD00000648
Boiling Pt:109…110 °C (760 mmHg)
Density:1.13 g/cm³
Molecular Weight:120.54 g/mol
Flash Pt:31 °C
EINECS:220-916-6
UN:1722
ADR:6.1
:GHS02
:GHS05
:GHS06
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4210 to 1.4240 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results