Tubing connectors, cross-shaped, BOLA
Supplier: Bohlender
BOLA
BOHLD573-08EA 42 EUR
BOHLD573-08
Tubing connectors, cross-shaped, BOLA
Connectors Connectors, Tubing
The Cross-shaped fitting, made of PTFE with four connectors for elastic tubing.
  • Universal chemical resistance
  • Temperature resistance from -200 up to 250 °C
  • Vacuum: suitable