Cesium chloride ≥99%, GPR RECTAPUR®
Specification Test Results
Assay | Min. 99 % |
Heavy metals (as Pb) | Max. 20 ppm |
SO4 (Sulphate) | Max. 100 ppm |
Fe (Iron) | Max. 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...