DNA markers, 100 bp
Supplier: Omega Bio-Tek
M01-01EA 0 EUR
OMEGM01-01
DNA markers, 100 bp
The 100 bp DNA ladder is ideal for determining the size of double-stranded DNA from 100 to 1000 base pairs. The ladder consists of fragments that range in size from 100 bp to 1000 bp in 100 bp increments. The 500 bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 500 bp are 30 ng. The 500 bp fragment is 60 ng. This ladder is pre-mixed with loading dye and is ready to use.