Calcium chloride cylinders, DURAN®
Supplier: DWK Life Sciences
SCOT215704207EA 0 EUR
SCOT215704207
Calcium chloride cylinders, DURAN®
DURAN®, borosilicat glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes