Blancs for reaction vessels, DURAN®
Supplier: DWK Life Sciences
SCOT210207105EA 111.01 EUR
SCOT210207105 SCOT210207602 SCOT210206306 SCOT210208607 SCOT210205404
Blancs for reaction vessels, DURAN®
Reaction Vessels Reaction Vessels, Flask Shape
DURAN® borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Round bottom cylinders.