Order Entry
Draining racks, with drip pan
Draining racks, with drip pan
Supplier:  BOCHEM
Certificates
undefined
Draining racks, with drip pan
Supplier:  BOCHEM
Product Information
  • Catalog #
    Supplier #
    Seq
    W×D×H
    Pk
    Availability
    Price
  • Draining rack, with 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm)
    Draining rack, with 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm)
    BOCH10140
    10140
    01
    420×160×610 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 182,00 €
    182,00 €
    New Customer Promotional Price
    1 * 1 items
       182,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Draining rack, wth 60 rods (95 mm) and 5 arches (2×110 mm and 3×70 mm)
    Draining rack, wth 60 rods (95 mm) and 5 arches (2×110 mm and 3×70 mm)
    BOCH10130
    10130
    02
    420×160×610 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 152,00 €
    152,00 €
    New Customer Promotional Price
    1 * 1 items
       152,00 €
    Specifications:
    Specifications
    Environmentally Preferable