Order Entry
Graduated pipettes, type 2, Class AS, works certified, MBL
undefined
Graduated pipettes, type 2, Class AS, works certified, MBL
  •  
    Capacity
    Division
    Tolerance
    Colour code
    Pk
  • 612-3964
    PRA828ASWC
    25 ml
    0,20 ml
    0,100 ± ml
    Green
    2
    1 * 2 items
    1 * 2 items - 31,70 €
    31,70 €
    1 * 2 items
       31,70 €
    Specifications
    Environmentally Preferable
  • 612-3961
    PRA822ASWC
    2 ml
    0,02 ml
    0,010 ± ml
    Black
    2
    1 * 2 items
    1 * 2 items - 21,60 €
    21,60 €
    1 * 2 items
       21,60 €
    Specifications
    Environmentally Preferable
  • 612-3960
    PRA820ASWC
    1 ml
    0,01 ml
    0,007 ± ml
    Yellow
    2
    1 * 2 items
    1 * 2 items - 21,40 €
    21,40 €
    1 * 2 items
       21,40 €
    Specifications
    Environmentally Preferable
  • 612-3963
    PRA826ASWC
    10 ml
    0,10 ml
    0,050 ± ml
    Orange
    2
    1 * 2 items
    1 * 2 items - 22,40 €
    22,40 €
    1 * 2 items
       22,40 €
    Specifications
    Environmentally Preferable
  • 612-3962
    PRA824ASWC
    5 ml
    0,05 ml
    0,030 ± ml
    Red
    2
    1 * 2 items
    1 * 2 items - 21,70 €
    21,70 €
    1 * 2 items
       21,70 €