You Searched For: Foodservice Supplies
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GLASWARENFABRIK KARL HECHT
Description: Reusable drinking straws are made from borosilicate glass 3.3.
Supplier: Apollo Scientific
Description: A mixture of natural coconut puree and maltodextrin. Use at a concentration of 15 to 30 g/L.
Catalog Number: (APOSBIPB852-100G)
Supplier: Apollo Scientific
Description: A mixture of natural banana puree and maltodextrin. Use at a concentration of 30 to 50 g/L.
UOM: 1 * 100 g
Supplier: HUPFER METALLWERKE
Description: Stainless steel 18/10.
Supplier: SEMADENI
Description: PP, translucent or white.
Supplier: CONTACTO
Description: Baking dish made of stainless steel 18/10, with matt polished finish.
Catalog Number: (CONC5405/300)
Supplier: CONTACTO
Description: ¹⁸/₁₀ stainless steel.
UOM: 1 * 1 items
Catalog Number: (213-1098)
Supplier: NEOLAB
Description: These paper cups feature a PE coating that prevents moisture penetration of the cup with moisture or humidity.
UOM: 1 * 100 items
Catalog Number: (LAMA2147)
Supplier: LAMAPLAST
Description: Made of HDPE, white.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom