Bottle top tilt measures, without bottle
Supplier: Hirschmann Laborgeräte
HIRS300016060EA 0 EUR
HIRS300016060
Bottle top tilt measures, without bottle
Bottle Top Tilt Measures
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Automatic dispenser particularly suited for dispensing suspensions.
These tilt measures with ground joint cone NS 29/32 can fit on all bottles/flasks with NS29/32 sockets.
Informace pro objednávání: Reservoir bottles available on request.