Reverse flow Cannon-Fenske viscometers for manual measurements
Supplier: SI Analytics
SCOI285403034EA 354 EUR
SCOI285403034 SCOI285403067 SCOI285403001 SCOI285403042 SCOI285403075 SCOI285403083 SCOI285403091 SCOI285403018 SCOI285403104 SCOI285403059 SCOI285403026 SCOI285403112
Reverse flow Cannon-Fenske viscometers for manual measurements
Viscometers Capillary Viscometers
Reverse flow viscometers calibrated with constant.
Suitable for opaque liquids manual measurements.