Pycnometer heads with/without bottle
Supplier: Lenz
LENZ05660006EA 85.74 EUR
LENZ05660006 LENZ05660007 LENZ05660206 LENZ05660207
Pycnometer heads with/without bottle
Specific Gravity Bottles
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes