Hygiene mask, 600
Supplier: 3M
MMMA600AEA 29.7 EUR
MMMA600A
Hygiene mask, 600
Masks Hygiene Masks
The 600 hygiene face mask provides lightweight and hygienic protection against a particular application, process or product from the wearer.
  • Moulded, green coloured, shell design
  • Metal nose-clip allows wearer to gain a good fit with the mask
  • Can also be used as a surgical mask for industrial and non medical processes
For use in applications such as hygienic food preparation, where no actual respiratory protection is required. The mask acts as a barrier between the food and the wearer, protecting the product from the wearer's exhaled breath.
Balení: 50 face masks per case.