Reusable earplugs, E-A-R™ UltraTech™
Supplier: 3M
MMMAUT01003EA 41.6 EUR
MMMAUT01003
Reusable earplugs, E-A-R™ UltraTech™
E-A-R™ Ultratech earplugs greatly improve the ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively.
  • Moisture resistant, washable and reusable plugs
  • Pre-moulded plugs made from flexible materials that fit easily to the ear
  • Compatible with other PPE
They provide flat attenuation across the hearing frequencies, making them ideal for musicians.
Signal-to-noise ratio: 21 dB