VWR® Disposal Bags
Supplier: VWR
VWR®
129-0584EA 502 EUR
129-0584 129-0585 129-0582 129-0583 129-0580 129-0581 129-0579 129-0588 129-0578 129-0589 129-0586 129-0587
VWR® Disposal Bags
Bags Waste Disposal Bags
For safe disposal of laboratory waste such as pipette tips, disposable Petri dishes, sample tubes, etc. PP, 40 µm thick, clear, with or without 'Biohazard' printing.
  • Heat resistant up to 145 °C
  • Autoclavable