You Searched For: Jerricans
32  results were found
SearchResultCount:"32"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: HDPE, opaque with PP screw caps.
Catalog Number: (216-1917)
Supplier: VWR Collection
Description: Disposable jerrican, made of HDPE, with integrated handle for easy carrying and pouring.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: Jerricans, Lowboys™, Nalgene®, flat form, , with screw cap and stopcock, PP, Capacity: 15 l, W×D×H: 368×368×203 mm, Colour: Natural, Neck I-Ø: 44 mm, Thread: 63 mm
Supplier: Thermo Fisher Scientific
Description: HDPE, translucent with white PP screw cap.
Supplier: Thermo Fisher Scientific
Description: HDPE, translucent with PP screw cap.
Catalog Number: (216-4402)
Supplier: Thermo Fisher Scientific
Description: HDPE, translucent with white PP closure.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: HDPE, fluorinated with fluorinated PP screw cap.
Supplier: DÃœPERTHAL GMBH & CO
Description: PE, electrostatically conductive.
Supplier: BOCHEM
Description: Stainless steel 18/10.
Supplier: BÃœRKLE
Description: HDPE, black, anti-static.
Supplier: BÃœRKLE
Description: HDPE, black (eletrically conductive).
Supplier: Kautex
Description: HDPE, natural colour.
Supplier: BÃœRKLE
Description: PP, translucent.
Supplier: BÃœRKLE
Description: HDPE, translucent or blue, with tamper evident screw cap.
Supplier: BÃœRKLE
Description: The filling support makes it easy to dispense liquids and correctly dose from Bürkle's 10 litre compact jerrycan.
Supplier: Corning
Description: HDPE, translucent.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 32
no targeter for Bottom