You Searched For: Filter Plates
37  results were found
SearchResultCount:"37"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Thermo Fisher Scientific
Description: Designed specifically for high throughput sample processing, these filter plates are suitable for vacuum filtration, centrifugation or positive pressu...
Supplier: Cytiva (Formerly Pall Lab)
Description: The AcroPrepâ„¢ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide r...
Sale
Supplier: Cytiva (Formerly Pall Lab)
Description: The AcroPrepâ„¢ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide r...
Supplier: Cytiva (Formerly Pall Lab)
Description: The Acroprep 24-well filter plate utilises Pall’s proprietary high performance multi-layer filter media and membranes to offer time savings, strong pe...
Sale
Supplier: Cytiva (Formerly Pall Lab)
Description: The AcroPrepâ„¢ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide r...
Supplier: Cytiva (Formerly Pall Lab)
Description: Ideaal voor high-throughput monstervoorbereidings- en detectieprocedures. AcroPrepâ„¢ filterplaten zijn vervaardigd uit chemisch bestendig, biologisch i...
Supplier: OMEGA BIO-TEK
Description: E-Z® 96 RNA plates (10 x 96), E-Z 96®, No. of wells: 96
Supplier: Cytiva (Formerly Pall Lab)
Description: The AcroPrepâ„¢ Advance filter plate portfolio includes a variety of membrane types and pore sizes that provide the best possible performance for a wide...
Supplier: Cytiva (Formerly Pall Lab)
Description: These plates are constructed from chemically resistant, biologically-inert polypropylene. Available in a variety of membrane configurations with a ser...
Supplier: Thermo Fisher Scientific
Description: Pierceâ„¢ protein precipitation plates provide an easy, 96-well format for simple, rapid and automatable precipitation and removal of proteins from plas...
Supplier: Cytiva (Formerly Pall Lab)
Description: The AcroPrepâ„¢ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide r...
Supplier: Cytiva (Formerly Pall Lab)
Description: The AcroPrepâ„¢ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide r...
Catalog Number: (738-0209)
Supplier: Cytiva (Formerly Pall Lab)
Description: The Pall AcroPrepâ„¢ Advance 96-well long tip filter plate for nucleic acid binding (NAB) incorporates a silica-based quartz glass fibre media to allow ...
UOM: 1 * 5 items
Supplier: Cytiva (Formerly Pall Lab)
Description: The AcroPrepâ„¢ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide r...
Supplier: Whatman products (Cytiva)
Description: The 10 ml UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and ...
Supplier: OMEGA BIO-TEK
Description: For simple and rapid homogenisation of cell and tissue lysates.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 37
no targeter for Bottom