Order Entry
Prefiltration capsules, Mini Profile®
Prefiltration capsules, Mini Profile®
undefined
Prefiltration capsules, Mini Profile®
  • Catalog No:
  • PALL12070
  • PALL12071
  • PALL12072
  • Type:
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
  • Inlet/outlet:
  • 13 mm hose barb
  • 13 mm hose barb
  • 13 mm hose barb
  • Max. temperature:
  • 50 °C
  • 50 °C
  • 50 °C
  • Max. pressure:
  • 5,0 bar
  • 5,0 bar
  • 5,0 bar
Product Information
  •  
    Seq
    Type
    Pore size
    Filtration area
    Pk
  • PALL12070
    12070
    03
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    1,5 µm
    90 cm²
    3
    1 * 3 items
    1 * 3 items - 346,00 €
    346,00 €
    1 * 3 items
       346,00 €
    Specifications
    Type
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    Environmentally Preferable
    Inlet/outlet
    13 mm hose barb
    Max. temperature
    50 °C
    Max. pressure
    5,0 bar
  • PALL12071
    12071
    04
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    3 µm
    90 cm²
    3
    1 * 3 items
    1 * 3 items - 346,00 €
    346,00 €
    1 * 3 items
       346,00 €
    Specifications
    Type
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    Environmentally Preferable
    Inlet/outlet
    13 mm hose barb
    Max. temperature
    50 °C
    Max. pressure
    5,0 bar
  • PALL12072
    12072
    05
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    5 µm
    90 cm²
    3
    1 * 3 items
    1 * 3 items - 346,00 €
    346,00 €
    1 * 3 items
       346,00 €
    Specifications
    Type
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    Environmentally Preferable
    Inlet/outlet
    13 mm hose barb
    Max. temperature
    50 °C
    Max. pressure
    5,0 bar