Medical light, SNE 136
Supplier: Waldmann
139-0186EA 447.12 EUR
139-0186 139-0187
Medical light, SNE 136
Lights Examination Lights
For intelligent lightning comfort, the SNE-136 provides broad ambient light to work area and neutral light with a 4000 K bulb. A built-in parabolic louvre directs light precisely to work areas and eliminates glare. It is designed for applications in industry and trade.
  • Spring loaded arm
  • Easy adjustment to your position
  • Built-in electronic ballast for energy efficiency
IP 20 Class 1
Certifikace: IP 20 Class 1
Delivery information: Supplied with 1,5 m mains lead and shock-proof plug.