This weather station features a perpetual calendar up to year 2099 with date, 12-/24-hour selectable time with snooze alarm, moon phase, tide indicator, sunset and sunrise times, barometric pressure, animated weather forecast, maximum/minimum memory for hygro-thermometer. RF wireless hygro-thermometer links up to three remote sensors.
- Easy to read icons
- Pressure bar chart, history and trend indicator
- Indoor and outdoor thermo-hygrometer with trend indicator
W×H: 127×201 mm
Certifikace: Compliant with RoHS. Traceable to standard from NIST. Conforms to ISO/IEC 17025:2005.
Delivery information: Supplied with remote sensor incl. batteries and two AA batteries for the main unit.