You Searched For: Pure and Ultra Pure Water Systems
69  results were found
SearchResultCount:"69"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: This powerful, intuitive laboratory water purification system provides superior and consistent ultrapure Type I and EDI-based Type II water from tap w...
Sale
Supplier: VWR Collection
Description: This compact 'plug and play' lab water system is an ideal choice for users who need up to 20 litres of ultrapure Type I water or RO water (Type III) p...
Sale
Supplier: VWR Collection
Description: This compact system produces EDI-based Type II water directly from tap water. Quality of Type II water meets or exceeds water quality as defined by AS...
Sale
Supplier: VWR Collection
Description: This 'two in one' lab water system produces ultrapure (Type I) and RO (Type III) water from tap water. Quality of ultrapure water meets or exceeds AST...
Sale
Supplier: VWR Collection
Description: This powerful and intuitive laboratory water purification system produces Type I ultrapure water from RO, distilled or deionised water. The product wa...
Supplier: Merck
Description: The Simplicity® portable water purification system can be located anywhere in your laboratory. It requires no special installation, no water feed conn...
Supplier: Merck
Description: RiOsâ„¢ Essential systems are ideal for laboratories needing reliable, constant production of Reverse Osmosis, RO (Type 3) water from potable tap water....
Supplier: Merck
Description: Purifies up to 3000 L/day with integrated storage, distribution and remote monitoring.
Supplier: Merck
Description: Direct-Q® 3 systems have been developed for simple, immediate use. Ultrapure Type I water is suitable for preparing buffers for liquid chromatography ...
Supplier: Merck
Description: Synergy® ultrapure water systems are ideal for laboratories with access to purified water that needs a 'point of use' supply of high quality ultrapure...
Supplier: Merck
Description: Reliable water purification for results reproducibility and lab productivity.
Supplier: Merck
Description: An integrated ultrapure and pure water system that is intelligent, easy to use and environmentally friendly.
Supplier: Merck
Description: To deliver reliable, high quality scientific results, you require consistently high quality ultrapure water. The Milli-Q® EQ 7000 ultrapure water syst...
Energy Efficient
Sustainable Materials
Waste Reducing
Supplier: Merck
Description: Milli-Q® IQ Element water purification and dispensing unit is designed to fit into your trace analysis workflow. The Milli-Q® IQ Element unit, combine...
Supplier: Merck
Description: Elix® Essential delivers Type 2 water customised to fit a broad range of research areas. It is designed to be fed from potable water and is the ideal ...
Catalog Number: (MILLZRDSVCL3GE)
Supplier: Merck
Description: This system is a very compact solution designed for low volume analysers. It efficiently combines reverse osmosis (RO) technology with deionisation (D...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 69
no targeter for Bottom