Water stills, Aquatron®
Supplier: Cole Parmer
stuart®
171-1202EA 4600 EUR
171-1202 171-1204 171-1203
Water stills, Aquatron®
Water Purification Systems Stills
Aquatron® units have fully automatic operation direct from a potable water supply - alternatively, an optional water feed conversion kit enables connection to any pre-treated water source. The distillate storage Level control switch fits any collection reservoir. A double coil condenser and boiler system combined with a VYCOR® silica glass sheathed heating element provides exceptionally high quality pyrogen-free distillate (although distillate should be tested). These stills can also be used with the filter and deionisation units for which the water feed conversion kit (Cat. No. 171-1210) is required.
  • Fully automatic operation
  • May be connected to a potable or pre-treated (conversion kit required) water supply
  • Operates with low quality or low pressure water source
  • Automatic cut-out for water supply failure
  • BioCote® antimicrobial protection
pH, conductivity and resistivity are affected by temperature and the presence of absorbed carbon dioxide. Specifications are based on tests conducted at 20 °C with CO₂-free water.
Care is needed to obtain pyrogen-free water and the distillate must be tested before use.
Water still heating elements are excluded from our 3 year warranty and are covered for 1000 hours use.
Delivery information: Supplied with wall mounting bracket (except A4000D).