You Searched For: Stills
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Cole Parmer
Description: Get high purity water with all-glass construction.
Supplier: SciLabware
Description: The QWS4 will produce 4 litres/hour of pyrogen-free* distilled water from its reliable 3kW UK manufactured, chromium plated heating element.
Supplier: Cole Parmer
Description: Produces cold distilled water for immediate use.
Supplier: LAUDA
Description: Water still suitable for single distillation and for bench mounting.
UL Listed
Supplier: LAUDA
Description: Water still (fully automatic) for double distillation, suitable for both bench and wall mounting.
Supplier: Cole Parmer
Description: Save money while maintaining high performance.
Catalog Number: (HECH40225010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Complete water distilling apparatus with heater and directions for use.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom