You Searched For: Boiling Baths
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GRANT INSTRUMENTS LTD.
Description: Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by ...
Supplier: LAUDA
Description: Vaporisation water baths which are ideal for gentle steaming work. The working temperature range is 25 to 100 °C.
Supplier: GESTIGKEIT HARRY
Description: Can be universally employed as either an air, fluid or sandbath for heating many of the various receptactles in laboratory use, regardless of size or ...
Supplier: GESTIGKEIT HARRY
Description: Precision heating baths which can be used for fluid, oil, air or as a sandbath for continuous operation. They are very well suited for use with rotary...
Catalog Number: (HECH43180010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: The Thermostatic waterbath, CE certified is fully electronic wear-free temperature control with digital display. Temperature control with adjustable n...
UOM: 1 * 1 items
Supplier: SELECTA
Description: Double skinned AISI 310 stainless steel tank with an exterior case of AISI 304 stainless steel. The heating elements embedded in INCOLOY heat and corr...
Supplier: NICKEL ELECTRO
Description: Designed for applications that require continuous boiling. Baths have a corrosion resistant stainless steel working environment, with heater and safet...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom